Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 25 μS/cm at 25°C NIST Traceable, Reagents

Catalog No. 01336704
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-704 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-704 Supplier Reagents Holdings Llc Supplier No. C7000025500A
Add to Cart
Add to Cart

Conductivity Standard, 25 μS/cm at 25°C NIST Traceable, Standard, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Poly Bottle
Specific Gravity 0.9982
Physical Form Liquid
Technique Calibration of Conductivity Instrument
Color Colorless
pH 6.7 to 7.3
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Conductivity Standard, 25 μS/cm at 25°C NIST Traceable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.