Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard (300,000 μS/cm at 25°C), MilliporeSigma™ Supelco™

Catalog No. 111002562 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2562 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2562 Supplier MilliporeSigma Supelco Supplier No. 66337500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conductivity standard can generally be used for the calibration of conductivity sensors for conductivity measurements. The conductivity standard solution for conductivity measurements can be either sodium chloride or potassium chloride solution of a definite concentration.

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-66-3
Density 1.063 to 1.133 g/cm3 at 20°C
Concentration 10-25% KCl (T)
Type Analytical Standard
Quantity 500 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.