Learn More
Aqua Solutions CONDUCTIVITY STANDARD 50.UMHOS
SDP
Catalog No. NC9380370
Encompass_Preferred
Conductivity Standard Traceable to N.I.S.T.; Clear, colorless, odorless liquid; 500mL; B.P. 100 deg.C; Specific Gravity: 1
Catalog No. NC9380370 Supplier Aqua Solutions Supplier No. 6941500ML
Conductivity Standard Traceable to N.I.S.T.; Clear, colorless, odorless liquid; 500mL; B.P. 100 deg.C; Specific Gravity: 1