Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 500,000 μS/cm (500 mS/cm) at 25°C, Reagents

Catalog No. 01334967
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-967 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-967 Supplier Reagents Holdings Llc Supplier No. CH4050001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conductivity Standard, 500,000 μS/cm (500 mS/cm) at 25°C, Reagent, use for Conductivity meter calibration. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Conductivity Standard, 500,000 μS/cm (500 mS/cm) at 25°C
Packaging Poly Bottle
CAS 7732-18-5, 7647-01-0
Specific Gravity 1.02
Physical Form Liquid
Technique Calibration of Conductivity Instrument
Color Colorless
pH 0
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.