Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 70 μS/cm at 25°C (33.0 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. 22361432 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
223614-32 1 L
223614-16 500 mL
223614-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 223614-32 Supplier Ricca Chemical Company Supplier No. 2236.1432
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 33.0ppm as NaCl
CAS Min % 98.0, 0.0032
CAS Max % 100.0, 0.0034
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.