Learn More
Aqua Solutions CONDUCTIVITY STD 150UMH 500 ML
SDP
Supplier:  Aqua Solutions 6903500ML
Encompass_Preferred
Conductivity Standard 150.0 umhos @ 25 C N.I.S.T. Traceable (500 ml)
Catalog No. NC0951217