Learn More
Cole-Parmer Thermo Scientific 011007 Conductivity Standard 1413 uS, 5 X 60 ml
Catalog No. NC0114641 Shop All Cole-Parmer Products
Encompass
Orion conductivity calibration solutions are available in portable 5 x 60 mL bottles. Portable calibration solutions are easier to carry than larger bottles, minimize wasted solution, and provide more volume than pouches. These standard solutions work with any manufacturer's conductivity meters.
Catalog No. NC0114641 Supplier Cole-Parmer Supplier No. 1970491
Orion conductivity calibration solutions are available in portable 5 x 60 mL bottles. Portable calibration solutions are easier to carry than larger bottles, minimize wasted solution, and provide more volume than pouches. These standard solutions work with any manufacturer's conductivity meters.