Learn More
Sensorex Conductivity/TDS Sensor,ATC
Catalog No. 501897540
Encompass_Preferred
The new SensoLab Series TDS Measurement Kit includes everything you need to start measuring conductivity or total dissolved solids for research, plant cultivation, hydroponics, aquaculture, and other applications. Includes our CM1000 benchtop conductivity meter with a fully compatible TDS sensor and built in temperature probe. Intelligent auto ranging selects the best measurement scale based on each sample, eliminating the guesswork.
Catalog No. 50-189-7540 Supplier Sensorex Supplier No. CM1000
The new SensoLab Series TDS Measurement Kit includes everything you need to start measuring conductivity or total dissolved solids for research, plant cultivation, hydroponics, aquaculture, and other applications. Includes our CM1000 benchtop conductivity meter with a fully compatible TDS sensor and built in temperature probe. Intelligent auto ranging selects the best measurement scale based on each sample, eliminating the guesswork.