Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity/TDS Standard, 12,880 μmho/cm (12.88 mS/cm), Reagents

Catalog No. 01335489
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-489 500 mL
01-335-490 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-489 Supplier Reagents Holdings Llc Supplier No. CC912880500A
Only null left
Add to Cart
Add to Cart

Conductivity/TDS Standard, 12,880 μmho/cm (12.88 mS/cm), NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Conductivity/TDS Standard, 12,880 μmho/cm (12.88 mS/cm)
Packaging Poly Bottle
CAS 7732-18-5, 7447-40-7
Specific Gravity 1.0052
Physical Form Liquid
Molecular Formula KCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 5.4 to 8.6
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.