Learn More
Cole-Parmer Oakton Conductivity/Temp Probe, for 6 and 6+ Series Meters; K=1.0
Catalog No. NC9489396 Shop All Cole-Parmer Products
Encompass_Preferred
Replacement Conductivity/Temp probe, K=1.0 for Oakton 6 and 6+ series meters
Catalog No. NC9489396 Supplier Cole-Parmer Supplier No. 3560655
Replacement Conductivity/Temp probe, K=1.0 for Oakton 6 and 6+ series meters