Learn More
METTLER TOLEDO CONDUCTVY STD SOL 500ML 1KUS
Catalog No. NC2187469 Shop All METTLER TOLEDO Products
Encompass
Conductivity Standard 500ml 1KuSMFG: 5807800310,000uS/cm, 500mL, KCI
This item is not returnable. View return policy
Catalog No. NC2187469 Supplier METTLER TOLEDO Supplier No. 58078003
Conductivity Standard 500ml 1KuSMFG: 5807800310,000uS/cm, 500mL, KCI