Learn More
Cayman Chemical Conduritol B epoxide, Purity: ≥95%, CAS: 6090-95-5, 25 mg
SDP
Catalog No. 502973140
Encompass_Preferred
An irreversible inhibitor of GBA (IC50 = 9 μM); does not inhibit non-lysosomal glucosylceramidase or cytosolic β-glucosidase
Catalog No. 50-297-3140 Supplier Cayman Chemical Supplier No. 1521625 MG
An irreversible inhibitor of GBA (IC50 = 9 μM); does not inhibit non-lysosomal glucosylceramidase or cytosolic β-glucosidase