Learn More
Cayman Chemical Conduritol B epoxide, Purity: ≥95%, CAS: 6090-95-5, 50 mg
SDP
Catalog No. 502972500
Encompass_Preferred
An irreversible inhibitor of GBA (IC50 = 9 μM); does not inhibit non-lysosomal glucosylceramidase or cytosolic β-glucosidase
Catalog No. 50-297-2500 Supplier Cayman Chemical Supplier No. 1521650 MG
An irreversible inhibitor of GBA (IC50 = 9 μM); does not inhibit non-lysosomal glucosylceramidase or cytosolic β-glucosidase