Learn More
Enzo Life Sciences Conduritol B Epoxide (5 mg). CAS: 6090-95-5
SDP
Supplier:  Enzo Life Sciences BMLS1040005
Encompass_Preferred
A potent, irreversible inhibitor of various plant β-glucosidases and mammalian glucocerebrosidase. Has been employed in in vitro and animal models for Gaucher’s disease. It raises intracellular levels of glucosylceramide by inhibiting glucosylceramidase. Alternative name: D,L-1,2-Anhydro-myo-inositol. Purity: ≥96% (TLC). Solubility: Soluble in water (25mg/ml) or in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1731