Learn More
Medchemexpress LLC Conduritol B epoxide | 6090-95-5 | MFCD00077326 | 99.9% | 162.14 g/mol | C6H10O5 | 1 ML
SDP
Catalog No. 5000212301
Encompass_Preferred
Conduritol B epoxide is an irreversible covalent inhibitor of acid β-glucosidase (GCase) used to block GCase activity in biochemical and cellular studies. It is supplied in solid form and as a 10 mM solution in DMSO for convenient preparation and dosing in research assays.
  • Irreversible covalent inhibitor of acid β-glucosidase (GCase).
  • Useful for biochemical and cellular assays investigating lysosomal function.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • High reported purity and defined molecular weight for reproducible results.
  • Stable under recommended storage conditions to preserve activity.
Catalog No. 50-002-12301 Supplier Medchemexpress LLC Supplier No. HY10094410MM 1ML
Conduritol B epoxide is an irreversible covalent inhibitor of acid β-glucosidase (GCase) used to block GCase activity in biochemical and cellular studies. It is supplied in solid form and as a 10 mM solution in DMSO for convenient preparation and dosing in research assays.
  • Irreversible covalent inhibitor of acid β-glucosidase (GCase).
  • Useful for biochemical and cellular assays investigating lysosomal function.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • High reported purity and defined molecular weight for reproducible results.
  • Stable under recommended storage conditions to preserve activity.