Learn More
Medchemexpress LLC Conduritol b epoxide | 6090-95-5 | MFCD00077326 | 99.9% | 162.14 g/mol | C6H10O5 | 100 MG
SDP
Catalog No. 5000213588
Encompass_Preferred
Conduritol B epoxide is an irreversible, covalent inhibitor of acid β-glucosidase (GCase) used in biochemical and cellular research to inhibit glucocerebrosidase activity. Supplied as a purified solid, it is used in enzymology studies, cellular models of lysosomal storage disorders, and mechanism-of-action experiments involving glycosidase pathways.
  • Irreversible inhibitor of acid β-glucosidase (GCase).
  • High purity (~99.9%) suitable for biochemical and cellular assays.
  • Molecular weight 162.14 g/mol and formula C6H10O5 for accurate stoichiometry.
  • White to off-white solid, stable for laboratory storage.
  • Available in convenient laboratory pack sizes including 100 MG.
  • Datasheet, certificate of analysis, and safety data sheet available.
Catalog No. 50-002-13588 Supplier Medchemexpress LLC Supplier No. HY100944100MG
Conduritol B epoxide is an irreversible, covalent inhibitor of acid β-glucosidase (GCase) used in biochemical and cellular research to inhibit glucocerebrosidase activity. Supplied as a purified solid, it is used in enzymology studies, cellular models of lysosomal storage disorders, and mechanism-of-action experiments involving glycosidase pathways.
  • Irreversible inhibitor of acid β-glucosidase (GCase).
  • High purity (~99.9%) suitable for biochemical and cellular assays.
  • Molecular weight 162.14 g/mol and formula C6H10O5 for accurate stoichiometry.
  • White to off-white solid, stable for laboratory storage.
  • Available in convenient laboratory pack sizes including 100 MG.
  • Datasheet, certificate of analysis, and safety data sheet available.