Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Med Vet International Cone (5mm X 58mm), J0045AD6, Each

Catalog No. 5000581749
Encompass_Preferred

Cone (5mm X 58mm), J0045AD6, Each

Catalog No. 50-005-81749 Supplier Med Vet International Supplier No. JOR45AD6
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cone (5mm X 58mm), J0045AD6, Each

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.