Learn More
Vector Laboratories ARR3 / Cone Arrestin Antibody (C‑Terminus), 100 µL
SDP
Supplier:  Vector Laboratories LSC368677100
Encompass
Cone Arrestin antibody LS-C368677 is an unconjugated rabbit polyclonal antibody to human Cone Arrestin (ARR3 ). Validated for ICC, IF, IHC and WB.
Catalog No. NC1361661