Learn More
Glass Expansion Cone Extraction Tool for skimmer removal and installation. Shimadzu 2030 ICP-MS.
SDP
Catalog No. 502703042
Encompass_Preferred
Cone Extraction Tool for skimmer removal and installation. Compatible with Shimadzu 2030 ICP-MS. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3042 Supplier Glass Expansion Supplier No. 708031573
Cone Extraction Tool for skimmer removal and installation. Compatible with Shimadzu 2030 ICP-MS. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001