Learn More
Waters Corp Waters Corporation Cone Gas Nozzle, for Mass Spectrometry Instruments, Ion Block Assembly Component
Catalog No. NC1986407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Cone Gas Nozzle is designed for various Waters mass spectrometry instruments, including Xevo, SYNAPT, and Select Series Instruments, as well as SQD2 and Vion. It serves as an integral component of the Ion Block Assembly.
  • One unit per package.
  • Compatible with MS system types.
  • Designed for use with a wide range of mass spectrometry instruments, including Xevo, SYNAPT, and Select Series Instruments.
  • Replacement is as required.
Catalog No. NC1986407 Supplier Waters Corp Supplier No. 700004216
This replacement Cone Gas Nozzle is designed for various Waters mass spectrometry instruments, including Xevo, SYNAPT, and Select Series Instruments, as well as SQD2 and Vion. It serves as an integral component of the Ion Block Assembly.
  • One unit per package.
  • Compatible with MS system types.
  • Designed for use with a wide range of mass spectrometry instruments, including Xevo, SYNAPT, and Select Series Instruments.
  • Replacement is as required.