Learn More
Glass Expansion Nickel Skimmer Cone, Dry Plasma - Nu Plasma II/AttoM ES/MKII ES (325-294)
SDP
Supplier:  Glass Expansion NU1014BNI
Encompass_Preferred
Nickel Skimmer Cone, Dry Plasma (325-294) For Nu Instruments Plasma2, Plasma3, Attom ES, Vitesse TOF-ICP-MS, and MKII ES ICP-MS. Suitable for laser ablation applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7681