Learn More
Glass Expansion Nickel Sampler Cone, Dry Plasma - Nu Plasma II/AttoM ES - Nickel Dry Sampler with Copper Core (319-646)
SDP
Catalog No. 501907680
Encompass_Preferred
Nickel Sampler Cone with Copper Core, Dry Plasma (319-646) For Nu Instruments Plasma2, Plasma3, Attom ES and Vitesse TOF-ICP-MS. Suitable for laser ablation applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7680 Supplier Glass Expansion Supplier No. NU1011BNI
Nickel Sampler Cone with Copper Core, Dry Plasma (319-646) For Nu Instruments Plasma2, Plasma3, Attom ES and Vitesse TOF-ICP-MS. Suitable for laser ablation applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001