Learn More
Perkin Elmer US LLC Cone Removal Tool for NexION 300/350/1000/2000
Catalog No. 502104384
Encompass_Preferred
This cone removal tool is used with NexION 300, 350, 1000, and 2000 ICP-MS series instruments.
Catalog No. 50-210-4384 Supplier Perkin Elmer US LLC Supplier No. W1034694
This cone removal tool is used with NexION 300, 350, 1000, and 2000 ICP-MS series instruments.