Learn More
Accuform™ Cone Top Warning Sign
Catalog No. 19169599 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
AUTHORIZED PERSONNEL ONLY
STOP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-169-599 AUTHORIZED PERSONNEL ONLY
19-169-598 STOP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-169-599 Supplier Accuform™ Supplier No. FBC764
Add to Cart
Add to Cart
Accuform™ Cone Top Warning Signs identify temporary hazardous conditions, displaying messages quickly and easily
  • Simply slip sign into the top of any flexible traffic cone
  • 10 in. x 14 in. rugged, plastic
  • Same message printed on both sides
  • Printed with weather- and fade-resistant inks
Specifications
Displayed Text AUTHORIZED PERSONNEL ONLY
Dimensions (L x W) 10 x 14 in.
Material Plastic