Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conessine, Tocris Bioscience™

Catalog No. 240950
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-095-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-095-0 Supplier R&D Systems Supplier No. 2409/50
This item is not returnable. View return policy

Potent and selective H3 receptor antagonist

Potent and selective histamine H3 receptor antagonist (pKi values are 7.61 and 8.27 at rat and human H3 receptors respectively). A steroidal alkaloid that displays in vitro antiplasmodial activity (IC50 = 1.04 μM). Also has high affinity for α2C adrenoceptors (pKi = 7.98).

Chemical Identifiers

CAS 546-06-5
Molecular Formula C24H40N2
Molecular Weight (g/mol) 356.60
MDL Number MFCD00016752
InChI Key GPLGAQQQNWMVMM-MYAJQUOBSA-N
Synonym conessine, roquessine, neriine, conessinum, wrightine, conessin, unii-ez38j9bbdf, ez38j9bbdf, konessin, conessine inn
PubChem CID 441082
ChEBI CHEBI:27965
IUPAC Name (1R,2S,5S,6S,9R,12S,13R,16S)-N,N,6,7,13-pentamethyl-7-azapentacyclo[10.8.0.0²,⁹.0⁵,⁹.0¹³,¹⁸]icos-18-en-16-amine
SMILES C[C@H]1[C@H]2CC[C@H]3[C@@H]4CC=C5C[C@H](CC[C@]5(C)[C@H]4CC[C@]23CN1C)N(C)C

Specifications

Quantity 50 mg
Formula Weight 356.59
Percent Purity >98%
Chemical Name or Material Conessine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.