Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Air Systems International Confined Space Fan Kit, 8 in., Explosion-Proof, 1/3 HP, 115 VAC, 1 ea

Catalog No. 502756647
Encompass_Preferred

This 8 in. explosion-proof axial fan kit is designed for ventilation in hazardous confined spaces. It includes a 1/3 HP fan, a 15 ft. conductive duct, and a conductive canister, providing reliable airflow in challenging environments. The system features a durable, conductive polyethylene construction and is engineered for safety in locations requiring explosion-proof equipment.

  • Approved for hazardous location usage in Class I, Division 1, Groups C and D, and Class II, Division 1, Groups E, F, and G environments
  • Delivers a maximum flow rate of 987 cfm in free air
  • Equipped with a 115 VAC, 60 Hz motor with a sealed on/off switch
  • Includes a 25 ft. power cord with no plug provided for user-specific wiring
  • Certified to ATEX and IECEx standards, and CSA/UL approved
  • Operates at a speed of 3,250 RPM

Catalog No. 50-275-6647 Supplier Grainger Supplier No. 15E893
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 8 in. explosion-proof axial fan kit is designed for ventilation in hazardous confined spaces. It includes a 1/3 HP fan, a 15 ft. conductive duct, and a conductive canister, providing reliable airflow in challenging environments. The system features a durable, conductive polyethylene construction and is engineered for safety in locations requiring explosion-proof equipment.

  • Approved for hazardous location usage in Class I, Division 1, Groups C and D, and Class II, Division 1, Groups E, F, and G environments
  • Delivers a maximum flow rate of 987 cfm in free air
  • Equipped with a 115 VAC, 60 Hz motor with a sealed on/off switch
  • Includes a 25 ft. power cord with no plug provided for user-specific wiring
  • Certified to ATEX and IECEx standards, and CSA/UL approved
  • Operates at a speed of 3,250 RPM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.