Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Air Systems International Axial Explosion Proof Fan, 115 V AC, 8 in. Duct, 1/3 hp, 1 ea.

Catalog No. 502756648
Encompass_Preferred

This axial explosion-proof fan is designed for use in hazardous locations, providing reliable ventilation for confined spaces. The unit is rated for indoor and outdoor use in challenging industrial environments.

  • Explosion-proof construction suitable for Class I, Division 1, Groups C and D; Class II, Division 1, Groups E, F and G environments
  • Includes 25 ft. conductive duct and conductive canister
  • Operates at 115 V AC, 60 Hz, single phase
  • Maximum free air flow rate of 987 CFM
  • Equipped with a built-in sealed on/off switch
  • Includes a 10 ft. power cord without a plug, requiring field wiring to NEC requirements
  • UL and CSA approved with T4 temperature code

Catalog No. 50-275-6648 Supplier Grainger Supplier No. 15E894
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This axial explosion-proof fan is designed for use in hazardous locations, providing reliable ventilation for confined spaces. The unit is rated for indoor and outdoor use in challenging industrial environments.

  • Explosion-proof construction suitable for Class I, Division 1, Groups C and D; Class II, Division 1, Groups E, F and G environments
  • Includes 25 ft. conductive duct and conductive canister
  • Operates at 115 V AC, 60 Hz, single phase
  • Maximum free air flow rate of 987 CFM
  • Equipped with a built-in sealed on/off switch
  • Includes a 10 ft. power cord without a plug, requiring field wiring to NEC requirements
  • UL and CSA approved with T4 temperature code

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.