Learn More
Grainger Air Systems International Confined Space Fan Kit, 8 in., 1/3 hp, 115 V AC, Black
Catalog No. 502756560
Encompass_Preferred
This confined space fan kit is designed for efficient and reliable ventilation in hazardous locations. Built with a rugged, lightweight, conductive polyethylene housing, this axial fan system is engineered to deliver fresh air safely in demanding job site environments where gas and dust are present. The kit includes a comprehensive set of conductive components to ensure integrity and performance in industrial applications.
  • Explosion-proof fan rated for Class I, Division 1, Groups C and D; Class II, Division 1, Groups E, F, and G environments
  • Powerful 1/3 hp motor providing up to 987 CFM in free air
  • Includes 25 ft and 6 ft conductive ducting, conductive canister, conductive 90 degree elbow, conductive saddle vent, and universal mount
  • Features a 25 ft power cord and a sealed on/off switch
  • Certified to CSA C/US standards for safety and performance
  • Operates at 115 V AC, 60 Hz
Catalog No. 50-275-6560 Supplier Grainger Supplier No. 15E898
This confined space fan kit is designed for efficient and reliable ventilation in hazardous locations. Built with a rugged, lightweight, conductive polyethylene housing, this axial fan system is engineered to deliver fresh air safely in demanding job site environments where gas and dust are present. The kit includes a comprehensive set of conductive components to ensure integrity and performance in industrial applications.
  • Explosion-proof fan rated for Class I, Division 1, Groups C and D; Class II, Division 1, Groups E, F, and G environments
  • Powerful 1/3 hp motor providing up to 987 CFM in free air
  • Includes 25 ft and 6 ft conductive ducting, conductive canister, conductive 90 degree elbow, conductive saddle vent, and universal mount
  • Features a 25 ft power cord and a sealed on/off switch
  • Certified to CSA C/US standards for safety and performance
  • Operates at 115 V AC, 60 Hz