Learn More
Grainger Air Systems International Ventilation Kit, 8 in., 974 CFM, Polyethylene
Catalog No. 502755435
Encompass_Preferred
This 8 in. polyethylene ventilation kit provides efficient airflow for confined space applications in non-hazardous locations, including boiler work, manhole entry, and underground vault maintenance. The system features a rugged, lightweight design suitable for demanding job site conditions.
  • Includes axial fan, 6 ft. and 15 ft. ducting with canister, saddle vent, 90° elbow, and universal mount
  • Provides 974 CFM free air performance
  • Operates on 115 VAC, 60 Hz power
  • 1/3 HP fan motor
  • CSA C/US certified
  • CE registered
  • Certified for outdoor use in all weather conditions
Catalog No. 50-275-5435 Supplier Grainger Supplier No. 15E895
This 8 in. polyethylene ventilation kit provides efficient airflow for confined space applications in non-hazardous locations, including boiler work, manhole entry, and underground vault maintenance. The system features a rugged, lightweight design suitable for demanding job site conditions.
  • Includes axial fan, 6 ft. and 15 ft. ducting with canister, saddle vent, 90° elbow, and universal mount
  • Provides 974 CFM free air performance
  • Operates on 115 VAC, 60 Hz power
  • 1/3 HP fan motor
  • CSA C/US certified
  • CE registered
  • Certified for outdoor use in all weather conditions