Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Air Systems International Axial Fan, Red, 8 in., 1/3 HP, 115 V AC, 1 ea.

Catalog No. 502756457
Encompass_Preferred

This 8 in. (20 cm) axial fan is a rugged and lightweight unit designed to provide fresh air for confined space ventilation. Certified for outdoor use in all weather conditions, this unit is built with a durable polyethylene housing and is suitable for operation in non-hazardous locations.

  • Features a 1/3 HP motor and 115 V AC operation at 60 Hz
  • Delivers a maximum air flow of 974 CFM in free air
  • Independently certified by CSA for use in the U.S. and Canada
  • Includes a 10 ft. (3.1 m) power cord
  • Operates at a speed of 3,250 RPM
  • Constructed from plastic for a lightweight design

Catalog No. 50-275-6457 Supplier Grainger Supplier No. 15E886
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 8 in. (20 cm) axial fan is a rugged and lightweight unit designed to provide fresh air for confined space ventilation. Certified for outdoor use in all weather conditions, this unit is built with a durable polyethylene housing and is suitable for operation in non-hazardous locations.

  • Features a 1/3 HP motor and 115 V AC operation at 60 Hz
  • Delivers a maximum air flow of 974 CFM in free air
  • Independently certified by CSA for use in the U.S. and Canada
  • Includes a 10 ft. (3.1 m) power cord
  • Operates at a speed of 3,250 RPM
  • Constructed from plastic for a lightweight design

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.