Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Air Systems International Confined Space Fan, 8 in., 1/3 HP, 115 VAC, 1-Piece Set

Catalog No. 502754056
Encompass_Preferred

These axial fans and kits provide a rugged, lightweight, and economical solution for fresh air ventilation in confined spaces under any condition. Designed for durability and versatility, they are certified for outdoor use in non-hazardous locations and are suitable for a wide range of industrial applications including utility maintenance, tank cleaning, and construction.

  • Features 1/3 HP motor with 974 CFM free air delivery
  • Constructed from durable polyethylene housing
  • Includes 8 in. (20.3 cm) diameter fan with 15 ft. duct and canister
  • Offers 115 VAC operation at 60 Hz
  • Equipped with a 10 ft. (3.1 m) power cord
  • Certified CSA and CUS approved

Catalog No. 50-275-4056 Supplier Grainger Supplier No. 15E887
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These axial fans and kits provide a rugged, lightweight, and economical solution for fresh air ventilation in confined spaces under any condition. Designed for durability and versatility, they are certified for outdoor use in non-hazardous locations and are suitable for a wide range of industrial applications including utility maintenance, tank cleaning, and construction.

  • Features 1/3 HP motor with 974 CFM free air delivery
  • Constructed from durable polyethylene housing
  • Includes 8 in. (20.3 cm) diameter fan with 15 ft. duct and canister
  • Offers 115 VAC operation at 60 Hz
  • Equipped with a 10 ft. (3.1 m) power cord
  • Certified CSA and CUS approved

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.