Learn More
Cell Signaling Technology TrueBlack™ Lipofuscin Autofluorescence Quencher, 1 mL
SDP
Catalog No. NC2309325
Encompass
TrueBlack™ Lipofuscin Autofluorescence Quencher is specifically formulated to reduce lipofuscin autofluorescence in aged human and animal tissue samples. It also minimizes general background fluorescence from sources including collagen, elastin, and red blood cells to enhance image clarity in immunofluorescence protocols. Supplied as a 20X concentrate, this reagent is designed for use in frozen tissue preparations.
  • Effectively quenches lipofuscin autofluorescence in tissues such as brain, heart, kidney, and retina
  • Reduces general background fluorescence
  • Supplied as a 20X concentrate in DMF
  • Stable for 12 months when stored at room temperature
  • Validated for compatibility with many common counterstains, such as DAPI
Catalog No. NC2309325 Supplier Cell Signaling Technology Supplier No. 92401S
TrueBlack™ Lipofuscin Autofluorescence Quencher is specifically formulated to reduce lipofuscin autofluorescence in aged human and animal tissue samples. It also minimizes general background fluorescence from sources including collagen, elastin, and red blood cells to enhance image clarity in immunofluorescence protocols. Supplied as a 20X concentrate, this reagent is designed for use in frozen tissue preparations.
  • Effectively quenches lipofuscin autofluorescence in tissues such as brain, heart, kidney, and retina
  • Reduces general background fluorescence
  • Supplied as a 20X concentrate in DMF
  • Stable for 12 months when stored at room temperature
  • Validated for compatibility with many common counterstains, such as DAPI