Learn More
Cayman Chemical Conglobatin, Purity: ≥85%, CAS: 72263-05-9, 2.5 mg
SDP
Catalog No. 502986300
Encompass_Preferred
An Hsp90 inhibitor; disrupts Hsp90 and Cdc37 binding; inhibits cell proliferation in HER2+ breast cancer cells (IC50 = 12.9-61.5 µM, respectively); decreases the number of invasive cells in vitro; inhibits cell proliferation in KYSE510 cells with an IC50 value of 9.31 µM; inhibits β integrin
Catalog No. 50-298-6300 Supplier Cayman Chemical Supplier No. 217662.5 MG
An Hsp90 inhibitor; disrupts Hsp90 and Cdc37 binding; inhibits cell proliferation in HER2+ breast cancer cells (IC50 = 12.9-61.5 µM, respectively); decreases the number of invasive cells in vitro; inhibits cell proliferation in KYSE510 cells with an IC50 value of 9.31 µM; inhibits β integrin