Learn More
Medchemexpress LLC HY-D0236 1g Medchemexpress, Congo Red CAS:573-58-0 Purity:>98%
SDP
Catalog No. 501933161
Encompass_Preferred
Medchemexpress, HY-D0236 1g Congo Red CAS:573-58-0 Congo Red is an azo dye. Congo Red (CR) binding been used as a diagnostic test for the presence of amyloid in tissue sections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3161 Supplier Medchemexpress LLC Supplier No. HYD02361G
Medchemexpress, HY-D0236 1g Congo Red CAS:573-58-0 Congo Red is an azo dye. Congo Red (CR) binding been used as a diagnostic test for the presence of amyloid in tissue sections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.