Learn More
Bioworld Congo Red, 100 g
Catalog No. 50199198
Encompass_Preferred
Certified Grade Absorbance Maximum(nm)- 497-500 Dye Content(%)- >75.0 P-15/P+15- 0.99-1.04
Catalog No. 50-199-198 Supplier Bioworld Supplier No. 217700101
Certified Grade Absorbance Maximum(nm)- 497-500 Dye Content(%)- >75.0 P-15/P+15- 0.99-1.04