Learn More
Apexbio Technology LLC Congo Red 573-58-0 500mg
SDP
Catalog No. 502240553
Encompass_Preferred
Congo Red (CAS 573-58-0) is a small-molecule inhibitor targeting the vesicular glutamate transporter (VGlut) It is designed to modulate glutamate storage and release thereby influencing synaptic transmission Congo Red exerts its biological activity primarily through selective non-covalent interaction with -sheet-rich structures such as those found in amyloid fibrils Based on these properties Congo Red holds research potential in the detection and qualitative assessment of amyloid deposits in tissue sections characterization of amyloidosis-related samples plant and fungal cell wall amyloids and lipopolysaccharide staining in Gram-negative bacterial outer membranes for biomedical and microbiological studies
This item is not returnable. View return policy
Catalog No. 50-224-0553 Supplier Apexbio Technology LLC Supplier No. B7766500
Congo Red (CAS 573-58-0) is a small-molecule inhibitor targeting the vesicular glutamate transporter (VGlut) It is designed to modulate glutamate storage and release thereby influencing synaptic transmission Congo Red exerts its biological activity primarily through selective non-covalent interaction with -sheet-rich structures such as those found in amyloid fibrils Based on these properties Congo Red holds research potential in the detection and qualitative assessment of amyloid deposits in tissue sections characterization of amyloidosis-related samples plant and fungal cell wall amyloids and lipopolysaccharide staining in Gram-negative bacterial outer membranes for biomedical and microbiological studies