Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014699 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4699 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-4699 Supplier MilliporeSigma Supelco Supplier No. 7576825MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 573-58-0
Physical Form Neat
Linear Formula C32H22N6 Na2O6S2
MDL Number MFCD00004028
Melting Point >360°C
Synonym Congo Red 4 B; Cosmos Red; Cotton Red B; Cotton Red C; Direct Red 28; Direct Red R; Direct Red Y
Type Analytical Standard
Quantity 25 mg
Formula Weight 696.66
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.