Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red, MP Biomedicals™

Catalog No. ICN10509980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10509980 100 g
ICN15071105 5 g
ICN10509990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. ICN10509980 Supplier MP Biomedicals, Inc Supplier No. 0210509980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym Cotton Red, Benzo Congo Red
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Red
Quantity 100 g
Physical Form Powder
Formula Weight 696.7
Chemical Name or Material Congo red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.