Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red, Reagents

Catalog No. 01336769
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-769 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-769 Supplier Reagents Holdings Llc Supplier No. C610800010C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Congo Red, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.66
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym Direct Red 28, Color Index 22120
IUPAC Name disodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
SMILES [Na+].[Na+].NC1=C2C=CC=CC2=C(C=C1N=NC1=CC=C(C=C1)C1=CC=C(C=C1)N=NC1=CC(=C2C=CC=CC2=C1N)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.995
Color Brown to Brown/Red
Quantity 10 g
Physical Form Solid
Chemical Name or Material Congo Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.