Learn More
Crescent Chemical Co Inc CONGO RED RESEARCH GRADE
SDP
Catalog No. 50247783
Encompass_Preferred
Congo Red research grade Tested for use in histology. Indicator pH 3.0 - 5.2 blue-reddish orange. λmax. 496 nm. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 27215.01 CAS 573-58-0
This item is not returnable. View return policy
Catalog No. 50-247-783 Supplier Crescent Chemical Co Inc Supplier No. SE27215.01
Congo Red research grade Tested for use in histology. Indicator pH 3.0 - 5.2 blue-reddish orange. λmax. 496 nm. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 27215.01 CAS 573-58-0