Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Congo Red Stain
SDP

Catalog No. 22021385
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-385 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-385 Supplier StatLab Supplier No. STCRE100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Congo Red Stain

  • Congo red stain is a synthetic dye primarily used for the detection of amyloid deposits in tissues
  • It belongs to the azo dye family, characterized by the presence of azo groups (-N=N-) that link aromatic compounds, giving the dye its distinctive red color
  • A MasterTech Special Stain Kit Component

Specifications

Grade Histological
Color Red
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.