Learn More
Hach Company Conical Adapter, 12 mm Diameter, for KF Titrator
Supplier: Hach Company LZE183
Conical adapter with a 12 mm diameter designed for use with Karl Fischer (KF) titrators. It provides a conical connection for KF titration sample ports and is intended to fit accessories or sample vessels that require a 12 mm conical opening.
- 12 mm diameter.
- Designed for use with KF titrators.
- Provides a conical connection for KF titration sample ports.