Learn More
Bellco Glass, Inc. The conical tissue homogenizer is designed for use with homogenizing motors. Ideal for tissues such as muscle, heart and lung. The conical tissue homogenizer has a large area for most efficient grinding. The clearance between pestles is .004-.006". Ca
SDP
Catalog No. 501945173
Encompass_Preferred
The conical tissue homogenizer is designed for use with homogenizing motors. Ideal for tissues such as muscle, heart and lung. The conical tissue homogenizer has a large area for most efficient grinding. The clearance between pestles is .004-.006". Ca
Catalog No. 50-194-5173 Supplier Bellco Glass, Inc. Supplier No. 197610021
The conical tissue homogenizer is designed for use with homogenizing motors. Ideal for tissues such as muscle, heart and lung. The conical tissue homogenizer has a large area for most efficient grinding. The clearance between pestles is .004-.006". Ca