Learn More
Bellco Glass, Inc. The conical tissue homogenizer is designed for use with homogenizing motors. Ideal for tissues such as muscle, heart and lung. The conical tissue homogenizer has a large area for most efficient grinding. The clearance between pestles is .004-.006". Ca
SDP
Supplier:  Bellco Glass, Inc. 197610025
Encompass_Preferred
The conical tissue homogenizer is designed for use with homogenizing motors. Ideal for tissues such as muscle, heart and lung. The conical tissue homogenizer has a large area for most efficient grinding. The clearance between pestles is .004-.006". Ca
Catalog No. 50-194-5177