Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bal Supply Conical Safety Bulb Pipette Filler
SCI_ED

Catalog No. S81083 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Includes:
Elastic band
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S81083 Elastic band
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81083 Supplier Bal Supply Supplier No. 5020PB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conical Safety Bulb Pipette Filler for easy use with all pipette sizes.

  • Included handy elastic band
  • Can be used with all sizes of pipettes

Specifications

Color Red
Material Rubber, Plastic, Silicone
Type Safety Bulb
For Use With (Equipment) All pipette sizes
Includes Elastic band
Dimensions (L x W x H) 75 x 25 x 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.