Learn More
Grainger Conical Spray Nozzle 2180 1/8 NPT Blue
Catalog No. 502764238
Encompass_Preferred
The Grainger Approved Conical Spray Nozzle is a brass nozzle with a 1/4" NPT female inlet and a conical spray pattern. It has a flow rate of 1.3 GPM at 40 PSI. The spray angle is 65 degrees and the orifice size is 0.09". It is made of brass for durability and has a maximum pressure rating of 100 PSI. The nozzle has a hex head for easy tightening and its compact size allows it to be used in tight spaces. This nozzle can be used for a variety of spraying applications.
Catalog No. 50-276-4238 Supplier Grainger Supplier No. 170J70
The Grainger Approved Conical Spray Nozzle is a brass nozzle with a 1/4" NPT female inlet and a conical spray pattern. It has a flow rate of 1.3 GPM at 40 PSI. The spray angle is 65 degrees and the orifice size is 0.09". It is made of brass for durability and has a maximum pressure rating of 100 PSI. The nozzle has a hex head for easy tightening and its compact size allows it to be used in tight spaces. This nozzle can be used for a variety of spraying applications.