Learn More
Research Products International Corp Conical Shaped Tissue Grinder, 6 per Bag
SDP
Catalog No. 50212946
Encompass
Conical-shaped tissue grinder has cone that tapers from 8.2mm to 2.9mm with a rounded tip. Overall length is 21cm. Steam autoclavable to 250°F (121°C). Packaged 6 per case.
Catalog No. 50-212-946 Supplier Research Products International Corp Supplier No. F199210001
Conical-shaped tissue grinder has cone that tapers from 8.2mm to 2.9mm with a rounded tip. Overall length is 21cm. Steam autoclavable to 250°F (121°C). Packaged 6 per case.