Learn More
MTC Bio 15ml PP (17x118mm), flat screw cap, 50/foam rack in sterile bag, 500/cs
Catalog No. 501127178
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Conical 15ml and 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Cat. No. C2605 features a skirted bottom that allows the tube to stand upright without a separate test tube rack. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.
Catalog No. 50-112-7178 Supplier MTC Bio Supplier No. C2600
This item has a minimum qty of 2 per supplier requirements.
Conical 15ml and 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Cat. No. C2605 features a skirted bottom that allows the tube to stand upright without a separate test tube rack. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.