Learn More
MTC Bio 50ml PP (29x115mm), flat screw cap, 25/foam rack in sterile bag, 500/cs
Catalog No. 501127181
Encompass_Preferred
Conical 15ml and 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Cat. No. C2605 features a ‘skirted’ bottom that allows the tube to stand upright without a separate test tube rack. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.
Catalog No. 50-112-7181 Supplier MTC Bio Supplier No. C2602
Conical 15ml and 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Cat. No. C2605 features a ‘skirted’ bottom that allows the tube to stand upright without a separate test tube rack. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.