Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Conical Tube Rack Holder
SDP

Catalog No. 22022583
Change view
Click to view available options
Mounting:
Magnet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-022-583 Magnet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-583 Supplier Poltex Supplier No. CONICRCKM
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poltex conical tube rack holder is a convenient holder for the Conical Tube Holder (CONIC) line of racks.

  • Convenient holder for the Conical Tube Holder (CONIC) line of racks
  • Holds up to 5 CONIC racks
  • Durable and cleanable PETG construction
  • Magnet mount (powerful magnets included), Tape mount (VHB Tape Included), or Wall mount (screws/anchors included)
  • Comes with rubber bumpons on base for optional countertop placement

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Tube Racks
Material Plastic
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
Mounting Magnet
Length (Metric) 101.60 mm
Product Type Tube Rack
Shelves None
Height (Metric) 266.70 mm
Width (Metric) 173.99 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.